Ethyl trans-β-methylcinnamate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl trans-beta-methylcinnamate, Ethyl (2E)-3-phenyl-2-butenoate

42800-0010 42800-0050
TS42800-0010EA 45 USD
TS42800-0010 TS42800-0050
Ethyl trans-β-methylcinnamate 97%
Ethyl trans-β-methylcinnamate
Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Boiling Pt: 142…144 °C (20hPa)
Density: 1.042 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00053762
CAS Number: 1504-72-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR