Toluene-3,4-dithiol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42118-0010
42118-0050
TS42118-0010EA
158
USD
TS42118-0010
TS42118-0050
Toluene-3,4-dithiol 95%
Toluene-3,4-dithiol
Formula:
C₇H₈S₂ Boiling Pt: 185 °C (84 mmHg) Melting Pt: 27 °C Density: 1.18 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00004844 CAS Number: 496-74-2 EINECS: 207-828-3 UN: 2810 Merck Index: 12,09669 |
Specification Test Results
Appearance | White to yellow Low melting crystalline solid |
Infrared spectrum | Conforms |
GC | ≥94 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...