Propylbenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

41843-0250 41843-1000 41843-5000
TS41843-0250EA 23 USD
TS41843-0250 TS41843-1000 TS41843-5000
Propylbenzene 98%
Propylbenzene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.86 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00009377
CAS Number: 103-65-1
EINECS: 203-132-9
UN: 2364
ADR: 3,III
Merck Index: 12,08029

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4900 to 1.4930 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +39° to +69° (20°C, 589 nm) (C=2, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR