Methyl-4-bromobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-bromobenzoate, 4-Bromobenzoic acid methyl ester

41407-0010 41407-0250
TS41407-0010EA 20 USD
TS41407-0010 TS41407-0250
Methyl-4-bromobenzoate 99%
Methyl-4-bromobenzoate
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 78…81 °C
MDL Number: MFCD00013531
CAS Number: 619-42-1
EINECS: 210-596-6
Merck Index: 13,01392

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 78 to 81 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR