Isopropyl salicylate 99%

Supplier: Thermo Scientific Chemicals
Warning

41289-1000 41289-2500
TS41289-1000EA 153 USD
TS41289-1000 TS41289-2500
Isopropyl salicylate 99%
Isopropyl salicylate
Formula: C₁₀H₁₂O₃
Boiling Pt: 122 °C (18 mmHg)
Density: 1.06 g/cm³
Flash Pt: >93 °C
Storage Temperature: Ambient
MDL Number: MFCD00035703
CAS Number: 607-85-2
EINECS: 210-143-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5110 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR