2-Iodophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

41248-0050 41248-0250 41248-1000
TS41248-0050EA 43 USD
TS41248-0050 TS41248-0250 TS41248-1000
2-Iodophenol 98%
2-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 186…187 °C (160 mmHg)
Melting Pt: 37…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013963
CAS Number: 533-58-4
EINECS: 208-569-9
Merck Index: 13,05057

Order Now

Specification Test Results

Appearance White to brown-red to brown Crystalline powder or low melting mass
Infrared spectrum Conforms
Melting point 37 to 43 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR