4-Hydroxyphthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

41213-0050 41213-0250
TS41213-0050EA 127 USD
TS41213-0050 TS41213-0250
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid
Formula: C₈H₆O₅
MW: 182.13 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00013984
CAS Number: 610-35-5
EINECS: 210-221-6

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR