(4-Hydroxyphenoxy)acetic acid 98%
Supplier: Thermo Scientific Chemicals
41209-0050
41209-0250
TS41209-0050EA
65
USD
TS41209-0050
TS41209-0250
(4-Hydroxyphenoxy)acetic acid 98%
(4-Hydroxyphenoxy)acetic acid
Formula:
(C₆H₄(OH))OCH₂COOH MW: 168.15 g/mol |
MDL Number:
MFCD00014362 CAS Number: 1878-84-8 |
Specification Test Results
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 152 to 156 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...