Ethyl phenylglyoxylate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl benzoylformate
41035-0050
41035-0250
TS41035-0050EA
29
USD
TS41035-0050
TS41035-0250
Ethyl phenylglyoxylate 95%
Ethyl phenylglyoxylate
Formula:
C₁₀H₁₀O₃ MW: 178.19 Boiling Pt: 138…139 °C (18 mmHg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009120 CAS Number: 1603-79-8 EINECS: 216-504-0 |
Specification Test Results
Appearance | Clear colorless to green Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5170 (20°C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...