4-Ethoxyphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

40967-0050 40967-0250 40967-1000
TS40967-0050EA 23 USD
TS40967-0050 TS40967-0250 TS40967-1000
4-Ethoxyphenol 99%
4-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 131 °C (9 mmHg)
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00002334
CAS Number: 622-62-8
EINECS: 210-748-1

Order Now

Specification Test Results

Appearance Beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR