trans,trans-Dibenzylideneacetone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

40900-0050 40900-0250 40900-1000
TS40900-0050EA 30 USD
TS40900-0050 TS40900-0250 TS40900-1000
trans,trans-Dibenzylideneacetone 98%
trans,trans-Dibenzylideneacetone
Formula: C₁₇H₁₄O
MW: 234.30 g/mol
Melting Pt: 107…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004790
CAS Number: 35225-79-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 108 to 114 °C
Assay ≥97.5 %
Pore volume 0.80 to 1.00 ml/g
Carbon (C) ≥16 %
Volatile matter ≤4 % (110 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR