Dinonyl phthalate mixture of isomers 96%
Supplier: Thermo Scientific Chemicals
40877-0250
40877-1000
TS40877-0250EA
108
USD
TS40877-0250
TS40877-1000
Dinonyl phthalate mixture of isomers 96%
Dinonyl phthalate
Formula:
C₂₆H₄₂O₄ Boiling Pt: 279…287 °C (760 mmHg) Density: 0.97 g/cm³ Flash Pt: 216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036237 CAS Number: 84-76-4 EINECS: 201-560-0 |
Specification Test Results
Appearance | Clear colorless to yellow-brown Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4820 to 1.4910 (20°C, 589 nm) |
GC | ≥95.0 % (sum of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...