O-Dianisidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine

40789-0050 40789-0250
TS40789-0050EA 36 USD
TS40789-0050 TS40789-0250
O-Dianisidine 97%
O-Dianisidine
Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00008372
CAS Number: 119-90-4
EINECS: 204-355-4

Order Now

Specification Test Results

Appearance Beige to pink to light brown to purple Crystalline powder or crystals (May darken on storage)
Infrared spectrum Conforms
Melting point 134 to 140 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR