1,3-Diiodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

40774-0050 40774-0250
TS40774-0050EA 34 USD
TS40774-0050 TS40774-0250
1,3-Diiodobenzene 98%
1,3-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Melting Pt: 33…37 °C
Density: 2.47 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00041731
CAS Number: 626-00-6
EINECS: 210-921-1

Order Now

Specification Test Results

Appearance Yellow to beige Powder or crystals or low melting crystalline mass
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR