Diethyl terephthalate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl benzene-1,4-dicarboxylate

40745-0250 40745-1000
TS40745-0250EA 85 USD
TS40745-0250 TS40745-1000
Diethyl terephthalate 98%
Diethyl terephthalate
Formula: C₁₂H₁₄O₄
Melting Pt: 42…45 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 43 to 47 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR