o-Cresol 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-2-methylbenzene, 2-Methylphenol, 2-Hydroxytoluene

40573-1000 40573-8000
TS40573-1000EA 34 USD
TS40573-1000 TS40573-8000
o-Cresol 99%, AcroSeal™
o-Cresol
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 191 °C (760 mmHg)
Melting Pt: 30…32 °C
Density: 1.04 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002226
CAS Number: 95-48-7
UN: 3455
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to amber Low melting solid or liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.2 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR