2-Chloronaphthalene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40484-0050
40484-0250
TS40484-0050EA
160
USD
TS40484-0050
TS40484-0250
2-Chloronaphthalene 99%
2-Chloronaphthalene
Formula:
C₁₀H₇Cl Boiling Pt: 256 °C (760 mmHg) Melting Pt: 57…60 °C Flash Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035731 CAS Number: 91-58-7 EINECS: 202-079-9 Merck Index: 13,02169 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 57 to 60 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...