Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate monohydrate 80% tech.

Supplier: Thermo Scientific Chemicals

Synonyms: 8-Amino-1-naphthol-3,6-disulphonic acid, monosodium salt monohydrate

40084-0050 40084-1000 40084-5000
TS40084-0050EA 26 USD
TS40084-0050 TS40084-1000 TS40084-5000
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulfonate monohydrate 80% tech.
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulphonate monohydrate
Formula: C₁₀H₈NNaO₇S₂·1H₂O
MW: 359.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150460
CAS Number: 312693-54-2

Order Now

Specification Test Results

Appearance Beige to grey Powder and/or lumps
Infrared spectrum Conforms
Loss on drying 4 % to 6 % (1 g, 150°C, 4 hrs)
HPLC ≥75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR