4'-Aminoacetanilide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(4-Aminophenyl)acetamide

40060-0250 40060-1000
TS40060-0250EA 50 USD
TS40060-0250 TS40060-1000
4'-Aminoacetanilide 95%
4'-Aminoacetanilide
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 267 °C
Melting Pt: 159…165 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007853
CAS Number: 122-80-5
EINECS: 204-576-6
Merck Index: 13,00410

Order Now

Specification Test Results

Appearance Pink to brown Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 165 °C
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR