4'-Methylacetanilide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide

40016-1000 40016-5000
TS40016-1000EA 56 USD
TS40016-1000 TS40016-5000
4'-Methylacetanilide 99%
4'-Methylacetanilide
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Boiling Pt: 307 °C (760 mmHg)
Melting Pt: 148…151 °C
Density: 1.21 g/cm³
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00008677
CAS Number: 103-89-9
EINECS: 203-155-4
Merck Index: 13,00076

Order Now

Specification Test Results

Appearance White to pink to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 147 to 151 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR