1-Acetylnaphthalene 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

40009-0010 40009-2500
TS40009-0010EA 296 USD
TS40009-0010 TS40009-2500
1-Acetylnaphthalene 95%
1-Acetylnaphthalene
Formula: C₁₂H₁₀O
Boiling Pt: 302 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.119 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004013
CAS Number: 941-98-0
EINECS: 213-384-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6270 to 1.6290 (20°C, 589 nm)
GC ≥94 %
Composition ≤2 % (CoCl2) Typical of fatty acids:


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR