4-Acetamidobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-(4-Formylphenyl)acetamide, 4'-Formylacetanilide

40002-0100 40002-0250 40002-1000
TS40002-0100EA 62 USD
TS40002-0100 TS40002-0250 TS40002-1000
4-Acetamidobenzaldehyde 98%
4-Acetamidobenzaldehyde
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00003380
CAS Number: 122-85-0
EINECS: 204-579-2

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR