4-Bromo-m-anisidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3-methoxyaniline, 5-Amino-2-bromoanisole, 4-Bromo-3-anisidine

39981-0010 39981-0050
TS39981-0010EA 42 USD
TS39981-0010 TS39981-0050
4-Bromo-m-anisidine 97%
4-Bromo-m-anisidine
Formula: C₇H₈BrNO
Melting Pt: 96…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664063
CAS Number: 19056-40-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 100 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR