((3-Bromopropoxy)methyl)benzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl 3-bromopropyl ether, 3-bromopropylbenzyl ether

39913-0010 39913-0100
TS39913-0010EA 43 USD
TS39913-0010 TS39913-0100
((3-Bromopropoxy)methyl)benzene 98%
((3-Bromopropoxy)methyl)benzene
Formula: C₁₀H₁₃BrO
MW: 229.117 g/mol
Boiling Pt: 130…132 °C (8 mmHg)
Density: 1.298 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134570
CAS Number: 54314-84-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5300 to 1.5330 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR