4-Fluoro-2-methoxyphenol 97%

Supplier: Thermo Scientific Chemicals

39899-0010 39899-0050
TS39899-0010EA 38 USD
TS39899-0010 TS39899-0050
4-Fluoro-2-methoxyphenol 97%
4-Fluoro-2-methoxyphenol

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR