5-Bromo-2-chloroanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

39890-0050 39890-0250
TS39890-0050EA 121 USD
TS39890-0050 TS39890-0250
5-Bromo-2-chloroanisole 98%
5-Bromo-2-chloroanisole
Formula: C₇H₆BrClO
MW: 221.48 g/mol
Boiling Pt: 240…245 °C (748 mmHg)
Melting Pt: 27…28 °C
Density: 1.631 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00078659
CAS Number: 16817-43-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR