N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Aminophenyl)carbamic Acid tert-Butyl Ester, 4-(tert-Butoxycarbonylamino)aniline, N-BOC-p-phenylenediamine, tert-Butyl (4-aminophenyl)carbamate, 4-(Boc-amino)aniline, N-Boc-1,4-phenylenediamine, tert-Butyl (4-Aminophenyl)carbamate

39833-0010 39833-0050 39833-0250
TS39833-0010EA 51 USD
TS39833-0010 TS39833-0050 TS39833-0250
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine 97%
N-(tert-Butoxycarbonyl)-1,4-phenylenediamine
Formula: C₁₁H₁₆N₂O₂
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00043022
CAS Number: 71026-66-9

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 115 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR