2,6-Dimethylbenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

39756-0010 39756-0050
TS39756-0010EA 39 USD
TS39756-0010 TS39756-0050
2,6-Dimethylbenzaldehyde 97%
2,6-Dimethylbenzaldehyde
Formula: C₉H₁₀O
Melting Pt: 27…30 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00128003
CAS Number: 1123-56-4

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR