3-Bromobenzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

39745-0050 39745-0250 39745-1000
TS39745-0050EA 33 USD
TS39745-0050 TS39745-0250 TS39745-1000
3-Bromobenzoyl chloride 97%
3-Bromobenzoyl chloride
Formula: C₇H₄BrClO
Boiling Pt: 74.5…75 °C (0.5 mmHg)
Density: 1.666 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5940 to 1.5960 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR