4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride
39704-0010
39704-0050
TS39704-0010EA
23
USD
TS39704-0010
TS39704-0050
4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride
Formula:
C₇H₈ClF₃N₂O MW: 228.60 g/mol Melting Pt: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053033 CAS Number: 133115-72-7 |
Specification Test Results
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...