2-Bromo-3-nitrotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39410-0010
39410-0050
TS39410-0010EA
20
USD
TS39410-0010
TS39410-0050
2-Bromo-3-nitrotoluene 98%
2-Bromo-3-nitrotoluene
Formula:
C₇H₆BrNO₂ Boiling Pt: 135…136 °C (8 mmHg) Melting Pt: 38…40 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134555 CAS Number: 41085-43-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow Crystalline powder or crystals or granules and/or chunks |
Infrared spectrum | Conforms |
Melting point | 36 to 40 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...