(4-(Benzyloxy)phenyl)hydrazine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39403-0010
39403-0050
TS39403-0010EA
74
USD
TS39403-0010
TS39403-0050
(4-(Benzyloxy)phenyl)hydrazine hydrochloride 98%
(4-(Benzyloxy)phenyl)hydrazine hydrochloride
Formula:
C₁₃H₁₅ClN₂O Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00137741 CAS Number: 52068-30-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Cream to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...