2-(Trifluoromethoxy)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane

39381-0010 39381-0050
TS39381-0010EA 58 USD
TS39381-0010 TS39381-0050
2-(Trifluoromethoxy)phenylboronic acid 97%
2-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01320763
CAS Number: 175676-65-0

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR