3,5-Dibromobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Danger

39315-0010 39315-0050 39315-0250
TS39315-0010EA 35 USD
TS39315-0010 TS39315-0050 TS39315-0250
3,5-Dibromobenzaldehyde 98+%
3,5-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
MW: 263.92
Storage Temperature: Refrigerator
MDL Number: MFCD00156887
CAS Number: 56990-02-4
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR