(4-Bromophenyl)acetic acid ethyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate

39299-0010 39299-0050 39299-0250
TS39299-0010EA 14 USD
TS39299-0010 TS39299-0050 TS39299-0250
(4-Bromophenyl)acetic acid ethyl ester 99%
(4-Bromophenyl)acetic acid ethyl ester
Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
Melting Pt: 29…31 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016333
CAS Number: 14062-25-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR