(4-(Bromomethyl)phenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Bromomethyl)phenylacetic acid

39290-0010 39290-0050
TS39290-0010EA 39 USD
TS39290-0010 TS39290-0050
(4-(Bromomethyl)phenyl)acetic acid 97%
(4-(Bromomethyl)phenyl)acetic acid
Formula: (C₆H₄(CH₂Br))CH₂COOH
MW: 229.07 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00010632
CAS Number: 13737-36-5
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR