α-Bromo-3-iodotoulene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide

39253-0010 39253-0050 39253-0250
TS39253-0010EA 41 USD
TS39253-0010 TS39253-0050 TS39253-0250
α-Bromo-3-iodotoulene 97%
α-Bromo-3-iodotoulene
Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 47…50 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00019016
CAS Number: 49617-83-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Light orange to red to brown Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 50 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR