3-Iodobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals

39207-0010 39207-0050
TS39207-0010EA 85 USD
TS39207-0010 TS39207-0050
3-Iodobenzaldehyde 99%
3-Iodobenzaldehyde
Formula: C₇H₅IO
MW: 232.02 g/mol
Melting Pt: 57…60 °C
MDL Number: MFCD00039573
CAS Number: 696-41-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR