4-Iodobenzaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

39206-0010 39206-0050
TS39206-0010EA 132 USD
TS39206-0010 TS39206-0050
4-Iodobenzaldehyde 97+%
4-Iodobenzaldehyde
Formula: C₇H₅IO
MW: 232.02 g/mol
Melting Pt: 77…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039576
CAS Number: 15164-44-0

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 75 to 80 °C
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR