4-(Bromomethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(Bromomethyl)phenyl)metaboric acid, (4-(Bromomethyl)phenyl)boronic acid, Dihydroxy(4-(bromomethyl)phenyl)borane, 4-Bromomethylphenylboronic acid, 4-(Bromomethyl)benzeneboronic acid

39157-0010 39157-0050
TS39157-0010EA 92 USD
TS39157-0010 TS39157-0050
4-(Bromomethyl)phenylboronic acid 97%
4-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
Melting Pt: 165…172 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01318113
CAS Number: 68162-47-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR