4-(Chloromethyl)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39115-0100
39115-0500
TS39115-0100EA
64
USD
TS39115-0100
TS39115-0500
4-(Chloromethyl)benzonitrile 98%
4-(Chloromethyl)benzonitrile
Formula:
C₈H₆ClN MW: 151.60 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019761 CAS Number: 874-86-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Off-white to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 80 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...