1,1-Diphenylethylene oxide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Diphenyloxirane

39103-0010 39103-0050
TS39103-0010EA 81 USD
TS39103-0010 TS39103-0050
1,1-Diphenylethylene oxide
1,1-Diphenylethylene oxide
Formula: C₁₄H₁₂O
MW: 196.249 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00040725
CAS Number: 882-59-7
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 52 to 57 °C
NMR ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR