4-tert-Butylbenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
p-tert-Butylbenzylamine
39101-0010
39101-0050
TS39101-0010EA
40
USD
TS39101-0010
TS39101-0050
4-tert-Butylbenzylamine 97%
4-tert-Butylbenzylamine
Formula:
C₁₁H₁₇N MW: 163.26 g/mol Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040754 CAS Number: 39895-55-1 UN: 2735 ADR: 8 III |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5220 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...