Bis(4-bromophenyl)acetylene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Bis(4-bromophenyl)ethyne

39094-0010 39094-0050 39094-0250
TS39094-0010EA 77 USD
TS39094-0010 TS39094-0050 TS39094-0250
Bis(4-bromophenyl)acetylene 97%
Bis(4-bromophenyl)acetylene
Formula: C₁₄H₈Br₂
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00185147
CAS Number: 2789-89-1

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 183 to 187 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR