Benzyl-2-bromoethyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
((2-Bromoethoxy)methyl)benzene, Benzyloxyethyl Bromide, (2-Bromoethoxymethyl)benzene
38887-0050
38887-0250
38887-1000
TS38887-0010EA
37.08
USD
TS38887-0010
TS38887-0050
TS38887-0250
TS38887-1000
Benzyl-2-bromoethyl ether 97%
Benzyl-2-bromoethyl ether
Formula:
C₉H₁₁BrO Boiling Pt: 254 °C (760 mmHg) Density: 1.36 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD01321307 CAS Number: 1462-37-9 UN: 3334 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5390 to 1.5420 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...