α-Bromo-4-nitro-o-cresol 95%

Supplier: Thermo Scientific Chemicals
Danger

38852-0050 38852-0250
TS38852-0050EA 48 USD
TS38852-0050 TS38852-0250
α-Bromo-4-nitro-o-cresol 95%
α-Bromo-4-nitro-o-cresol
Formula: C₇H₆BrNO₃
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007340
CAS Number: 772-33-8
EINECS: 212-248-9
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 149 °C
Titration Argentometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR