9-Benzhydrylidene-10-anthrone 95%

Supplier: Thermo Scientific Chemicals

38826-0010 38826-0050
TS38826-0010EA 16 USD
TS38826-0010 TS38826-0050
9-Benzhydrylidene-10-anthrone 95%
9-Benzhydrylidene-10-anthrone
Formula: C₂₇H₁₈O
Melting Pt: 203…207 °C
Storage Temperature: Ambient
CAS Number: 667-91-4

Order Now

Specification Test Results

Appearance Yellow to beige Powder
Infrared spectrum Conforms
Melting point 202 to 212 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR