Biphenyl-4,4'-dicarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid

38730-0050 38730-0250
TS38730-0050EA 99 USD
TS38730-0050 TS38730-0250
Biphenyl-4,4'-dicarboxylic acid 98%
Biphenyl-4,4'-dicarboxylic acid
Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002554
CAS Number: 787-70-2

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR