4-Acetoxy-3-methoxybenzaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Formyl-2-methoxyphenyl acetate, Vanillin acetate
38594-0050
38594-0250
38594-1000
TS38594-0050EA
29
USD
TS38594-0050
TS38594-0250
TS38594-1000
4-Acetoxy-3-methoxybenzaldehyde 96%
4-Acetoxy-3-methoxybenzaldehyde
Formula:
C₁₀H₁₀O₄ Melting Pt: 73…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003362 CAS Number: 881-68-5 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 73 to 80 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...