1-(2-Bromoethoxy)-3-nitrobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

38526-0250 385260050
TS38526-0250EA 144 USD
TS38526-0250 TS385260050
1-(2-Bromoethoxy)-3-nitrobenzene 97%
1-(2-Bromoethoxy)-3-nitrobenzene
Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Boiling Pt: 140…145 °C (2 mmHg)
Melting Pt: 39 °C
MDL Number: MFCD07783648
CAS Number: 13831-59-9

Order Now

Specification Test Results

Appearance Yellow-green to brown Low melting solid
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR